Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD98 Antibody (1C11.7E3), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NBP236491W Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP236491W 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP236491W Supplier Novus Biologicals Supplier No. NBP236491AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD98 Monoclonal specifically detects CD98 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen CD98
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1C11.7E3
Conjugate Alexa Fluor 405
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. P08195
Gene Alias 4F2, 4F2hc, 4T2HC, antigen identified by monoclonal antibodies 4F2, TRA1.10, TROP4, and T43,4F2HC, CD98, CD98 antigen, CD98 heavy chain, CD98HC, heavy chain, Lymphocyte activation antigen 4F2 large subunit, MDU1antigen defined by monoclonal 4F2, heavy chain, monoclonal 44D7, NACAE4F2 cell-surface antigen heavy chain, solute carrier family 3 (activators of dibasic and neutral amino acidtransport), member 2,4F2 heavy chain antigen
Gene Symbols SLC3A2
Host Species Mouse
Immunogen Partial recominant human CD98 (between amino acids 200-400) [UniProt P08195]
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, Immunology, Lipid and Metabolism, Plasma Membrane Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6520
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.