Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD98 Antibody (HBJ127), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB016290 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB016290 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB016290 Supplier Novus Biologicals Supplier No. NBP281109AF750
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CD98 Monoclonal specifically detects CD98 in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen CD98
Applications ELISA, Immunofluorescence
Classification Monoclonal
Clone HBJ127
Conjugate Alexa Fluor 750
Dilution ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias 4F2, 4F2hc, 4T2HC, antigen identified by monoclonal antibodies 4F2, TRA1.10, TROP4, and T43,4F2HC, CD98, CD98 antigen, CD98 heavy chain, CD98HC, heavy chain, Lymphocyte activation antigen 4F2 large subunit, MDU1antigen defined by monoclonal 4F2, heavy chain, monoclonal 44D7, NACAE4F2 cell-surface antigen heavy chain, solute carrier family 3 (activators of dibasic and neutral amino acidtransport), member 2,4F2 heavy chain antigen
Gene Symbols SLC3A2
Host Species Rabbit
Immunogen HBJ127 was obtained from hybridomas generated by the fusion of mouse myeloma cells and spleen cells from mice which had been immunized with T24 hyman bladder cancer cells.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cell Biology, Cellular Markers, Immunology, Lipid and Metabolism, Plasma Membrane Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6520
Test Specificity HBJ127 binds to the CD98 heavy chain at an epitope consisting of residues AFS (aa 442-444) (Itoh et al, 2007). The antibody has not been shown to cross-react with other species. CD98 is expressed at high levels on monocytes and at very low level on peripheral blood T and B lymphocytes, splenocytes, NK cells and granulocytes. CD98 is a disulfide-linked and glycosylated type II integral membrane protein. This protein has roles in normal and neoplastic cell growth and is required for the function of light chain amino acid transporters. It is also involved in sodium-independent transport of large neitral amino acids and in guiding and targeting LAT1 and LAT2 to the plasma membrane. When associated with SLC7A6 or SLC7A7, CD98 acts as an arginine/glutamine exchanger.
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.