Learn More
Description
Specifications
Specifications
| Antigen | CD98 |
| Applications | ELISA, Immunofluorescence |
| Classification | Monoclonal |
| Clone | HBJ127 |
| Conjugate | DyLight 488 |
| Dilution | ELISA, Immunocytochemistry/Immunofluorescence |
| Formulation | 50mM Sodium Borate |
| Gene Alias | 4F2, 4F2hc, 4T2HC, antigen identified by monoclonal antibodies 4F2, TRA1.10, TROP4, and T43,4F2HC, CD98, CD98 antigen, CD98 heavy chain, CD98HC, heavy chain, Lymphocyte activation antigen 4F2 large subunit, MDU1antigen defined by monoclonal 4F2, heavy chain, monoclonal 44D7, NACAE4F2 cell-surface antigen heavy chain, solute carrier family 3 (activators of dibasic and neutral amino acidtransport), member 2,4F2 heavy chain antigen |
| Gene Symbols | SLC3A2 |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
