Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD98 Rabbit anti-Mouse, Clone: RL388, Novus Biologicals™
SDP

Catalog No. NB153589 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.2 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB153589 0.025 mg
NB153590 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB153589 Supplier Novus Biologicals Supplier No. NBP3120360.025MG
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CD98 Monoclonal antibody specifically detects CD98 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation, Functional Assay

Specifications

Antigen CD98
Applications Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation, Functional Assay
Classification Monoclonal
Clone RL388
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Functional
Formulation PBS
Gene Alias 4F2, 4F2hc, 4T2HC, antigen identified by monoclonal antibodies 4F2, TRA1.10, TROP4, and T43,4F2HC, CD98, CD98 antigen, CD98 heavy chain, CD98HC, heavy chain, Lymphocyte activation antigen 4F2 large subunit, MDU1antigen defined by monoclonal 4F2, heavy chain, monoclonal 44D7, NACAE4F2 cell-surface antigen heavy chain, solute carrier family 3 (activators of dibasic and neutral amino acidtransport), member 2,4F2 heavy chain antigen
Host Species Rabbit
Immunogen This antibody was raised by immunising rats with an IL 2-producing subline of the mouse T lymphoma EL4.
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cell Biology, Cellular Markers, Immunology, Lipid and Metabolism, Plasma Membrane Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6520
Target Species Mouse
Content And Storage Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C.
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.