Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD98 Rabbit anti-Mouse, Clone: RL388, Novus Biologicals™

Rabbit Monoclonal Antibody
$268.00 - $584.50
Specifications
Antigen | CD98 |
---|---|
Clone | RL388 |
Concentration | 1 mg/mL |
Dilution | Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Functional |
Applications | Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation, Functional Assay |
Description
CD98 Monoclonal antibody specifically detects CD98 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation, Functional AssaySpecifications
CD98 | |
1 mg/mL | |
Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation, Functional Assay | |
Unconjugated | |
RUO | |
Mouse | |
4F2, 4F2hc, 4T2HC, antigen identified by monoclonal antibodies 4F2, TRA1.10, TROP4, and T43,4F2HC, CD98, CD98 antigen, CD98 heavy chain, CD98HC, heavy chain, Lymphocyte activation antigen 4F2 large subunit, MDU1antigen defined by monoclonal 4F2, heavy chain, monoclonal 44D7, NACAE4F2 cell-surface antigen heavy chain, solute carrier family 3 (activators of dibasic and neutral amino acidtransport), member 2,4F2 heavy chain antigen | |
This antibody was raised by immunising rats with an IL 2-producing subline of the mouse T lymphoma EL4. | |
Primary | |
Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
RL388 | |
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Functional | |
Monoclonal | |
Rabbit | |
Biologically Active Proteins, Cell Biology, Cellular Markers, Immunology, Lipid and Metabolism, Plasma Membrane Markers, Signal Transduction | |
PBS | |
6520 | |
IgG κ | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title