Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD99 Antibody (MIC2/3478R), Biotin, Novus Biologicals™
SDP

Catalog No. NB391887 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB391887 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB391887 Supplier Novus Biologicals Supplier No. NBP308589B
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CD99 Monoclonal antibody specifically detects CD99 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen CD99
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MIC2/3478R
Conjugate Biotin
Formulation PBS
Gene Alias 12E7, CD99 antigenY homolog, CD99 molecule, E2 antigen, HBA71, MIC2 (monoclonal 12E7), MIC2Y, MSK5X, Protein MIC2, surface antigen MIC2, T-cell surface glycoprotein E2
Host Species Rabbit
Immunogen Synthetic peptide corresponding to CD99 residues within aa85 C terminal of CD99 was used as an immunogen. (Uniprot: P14209 )
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 4267
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.