Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD99 Antibody (OTI1D5), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP271197A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP271197A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP271197A32 Supplier Novus Biologicals Supplier No. NBP271197AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD99 Monoclonal specifically detects CD99 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen CD99
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI1D5
Conjugate Alexa Fluor 532
Dilution Western Blot, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias 12E7, CD99 antigenY homolog, CD99 molecule, E2 antigen, HBA71, MIC2 (monoclonal 12E7), MIC2Y, MSK5X, Protein MIC2, surface antigen MIC2, T-cell surface glycoprotein E2
Gene Symbols CD99
Host Species Mouse
Immunogen Full length recombinant protein of human CD99 (NP_002405) produced in HEK293T cells.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 4267
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.