Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDAN1 Antibody (Cod177) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB395520 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB395520 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB395520 Supplier Novus Biologicals Supplier No. NBP2501760.025ML
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CDAN1 Monoclonal antibody specifically detects CDAN1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen CDAN1
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone Cod177
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500
Formulation PBS
Gene Alias CDA1, CDAI, CDA-I, codanin, codanin-1, congenital dyserythropoietic anemia, type I, discs lost homolog, DLT, PRO1295
Host Species Mouse
Immunogen aa 294-607 of cloned human protein
Purification Method Protein A purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology
Primary or Secondary Primary
Gene ID (Entrez) 146059
Target Species Human, Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.