Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDAN1 Antibody (Cod177) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$196.10
Specifications
Antigen | CDAN1 |
---|---|
Clone | Cod177 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:100 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500 |
Applications | Western Blot, Immunofluorescence |
Description
CDAN1 Monoclonal antibody specifically detects CDAN1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
CDAN1 | |
1 mg/mL | |
Western Blot, Immunofluorescence | |
Unconjugated | |
Mouse | |
Cardiovascular Biology | |
PBS | |
146059 | |
IgG2b | |
Protein A purified |
Cod177 | |
Western Blot 1:100 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
CDA1, CDAI, CDA-I, codanin, codanin-1, congenital dyserythropoietic anemia, type I, discs lost homolog, DLT, PRO1295 | |
aa 294-607 of cloned human protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title