Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDAN1 Antibody (Cod177), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP250176UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250176UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250176UV Supplier Novus Biologicals Supplier No. NBP250176UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CDAN1 Monoclonal specifically detects CDAN1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen CDAN1
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone Cod177
Conjugate DyLight 350
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CDA1, CDAI, CDA-I, codanin, codanin-1, congenital dyserythropoietic anemia, type I, discs lost homolog, DLT, PRO1295
Gene Symbols CDAN1
Host Species Mouse
Immunogen aa 294-607 of cloned human protein
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology
Primary or Secondary Primary
Gene ID (Entrez) 146059
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.