Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cdc13 Antibody (6F11/2), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB310272 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB310272 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB310272 Supplier Novus Biologicals Supplier No. NB200576AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cdc13 Monoclonal antibody specifically detects Cdc13 in S. pombe, Yeast samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen Cdc13
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 6F11/2
Conjugate Alexa Fluor 750
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate
Gene Alias Cell division control protein 13, G2/M B-type cyclin Cdc13, YDL220C
Host Species Mouse
Immunogen Full length Cdc13
Purification Method Protein A purified
Quantity 0.2 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species S. pombe, Yeast
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.