Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cdc13 Antibody (6F11/2), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB001126 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB001126 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB001126 Supplier Novus Biologicals Supplier No. NB200576JF5490.2ML
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cdc13 Monoclonal specifically detects Cdc13 in S. pombe, Yeast samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Cdc13
Applications Immunohistochemistry
Classification Monoclonal
Clone 6F11/2
Conjugate Janelia Fluor 549
Dilution Immunohistochemistry
Formulation 50 mM Sodium Borate
Gene Alias Cell division control protein 13, G2/M B-type cyclin Cdc13, YDL220C
Host Species Mouse
Immunogen Full length cdc13
Quantity 0.2 mL
Target Species S. pombe, Yeast
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.