Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDC2/CDK1 Antibody (POH-1; same as cdc2.1), DyLight 680, Novus Biologicals™
SDP

Catalog No. NBP234711FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP234711FR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP234711FR Supplier Novus Biologicals Supplier No. NBP234711FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CDC2/CDK1 Monoclonal specifically detects CDC2/CDK1 in Human, Bovine, Mink, Mustelid, Monkey, Drosophila (Negative), Mouse (Negative), Rat (Negative), Xenopus (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen CDC2/CDK1
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone POH-1; same as cdc2.1
Conjugate DyLight 680
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Gene Alias CDC28A, CDC2MGC111195, cell cycle controller CDC2, Cell division control protein 2 homolog, Cell division protein kinase 1, cyclin-dependent kinase 1, DKFZp686L20222, EC 2.7.11.22, EC 2.7.11.23, G1 to S and G2 to M, p34 protein kinase, P34CDC2
Gene Symbols CDK1
Host Species Mouse
Immunogen Recombinant full-length human p34cdc2 protein (Uniprot: P06493)
Molecular Weight of Antigen 34 kDa
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cell Cycle and Replication, Core ESC Like Genes, Mitotic Regulators, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 983
Test Specificity Recognizes a 34kDa protein, which is identified as cyclin dependent kinase 1 (cdk1) or p34cdc2 protein kinase. cdk1 p34cdc2 plays a crucial role during cell division and is most active during mitosis. It is predominantly localized in the nucleus. It is a serine/threonine kinase, which is activated by cyclin, presumably by de-phosphorylation of tyrosine residues. Activated cdk1 p34cdc2 performs specific functions during mitosis, including nuclear envelope breakdown and chromosome condensation.
Target Species Human, Bovine, Mustelid, Primate, Drosophila (Negative), Mouse (Negative), Rat (Negative), Xenopus (Negative)
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.