Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDC2/CDK1 Antibody (Y100.4), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB310374 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB310374 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB310374 Supplier Novus Biologicals Supplier No. NB1002716MFV500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CDC2/CDK1 Monoclonal antibody specifically detects CDC2/CDK1 in S. pombe samples. It is validated for Western Blot

Specifications

Antigen CDC2/CDK1
Applications Western Blot
Classification Monoclonal
Clone Y100.4
Conjugate mFluor Violet 500 SE
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias CDC28A, CDC2MGC111195, cell cycle controller CDC2, Cell division control protein 2 homolog, Cell division protein kinase 1, cyclin-dependent kinase 1, DKFZp686L20222, EC 2.7.11.22, EC 2.7.11.23, G1 to S and G2 to M, p34 protein kinase, P34CDC2
Host Species Mouse
Immunogen Peptide spanning the C-terminal 14 residues of S. pombe CDC2/CDK1
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cell Cycle and Replication, Core ESC Like Genes, Mitotic Regulators, Phospho Specific, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 983
Target Species S. pombe
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.