Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cdc23 Rabbit anti-Human, Mouse, Rat, Clone: 1Q2I3, Novus Biologicals™
SDP

Catalog No. NB168354 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB168354 20 μg
NB168353 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB168354 Supplier Novus Biologicals Supplier No. NBP31663120UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Cdc23 Monoclonal antibody specifically detects Cdc23 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence

Specifications

Antigen Cdc23
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone 1Q2I3
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias ANAPC8anaphase promoting complex subunit 8, Anaphase-promoting complex subunit 8, APC8CDC23 (cell division cycle 23, yeast, homolog), cell division cycle 23 homolog (S. cerevisiae), cell division cycle protein 23 homolog, CUT23, Cyclosome subunit 8
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 498-597 of human Cdc23 (Q9UJX2).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 8697
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.