Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDC25A Mouse anti-Human, Clone: DCS121, Novus Biologicals™
SDP

Catalog No. NB151717 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB151717 100 μg
NB151718 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB151717 Supplier Novus Biologicals Supplier No. NBP311882100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CDC25A Monoclonal antibody specifically detects CDC25A in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen CDC25A
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone DCS121
Conjugate Unconjugated
Dilution Western Blot 1 - 10 ug/ml, Immunoprecipitation 1 - 10 ug/ml
Formulation PBS
Gene Alias CDC25A2, CDC25A2-CAG isoform, cell division cycle 25 homolog A (S. cerevisiae), cell division cycle 25 homolog A (S. pombe), cell division cycle 25A, Dual specificity phosphatase Cdc25A, EC 3.1.3.48, M-phase inducer phosphatase 1
Host Species Mouse
Immunogen Mice were immunized with a purified recombinant CDC25A protein of 65kD protein (Uniprot: P30304)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cancer, Cell Cycle and Replication, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Immunology, MAP Kinase Signaling, Phospho Specific, Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 993
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.