Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDC25A Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB168943 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB168943 100 μg
NB168944 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB168943 Supplier Novus Biologicals Supplier No. NBP316932100UL
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CDC25A Polyclonal antibody specifically detects CDC25A in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen CDC25A
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 50% glycerol, pH7.3
Gene Alias CDC25A2, CDC25A2-CAG isoform, cell division cycle 25 homolog A (S. cerevisiae), cell division cycle 25 homolog A (S. pombe), cell division cycle 25A, Dual specificity phosphatase Cdc25A, EC 3.1.3.48, M-phase inducer phosphatase 1
Host Species Rabbit
Immunogen A synthetic phosphorylated peptide around S76 of human CDC25A (NP_001780.2).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cancer, Cell Cycle and Replication, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Immunology, MAP Kinase Signaling, Phospho Specific, Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 993
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.