Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDC25B Antibody (3D6), Novus Biologicals™
SDP

Catalog No. NB297633 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB297633 100 μL
NB297632 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB297633 Supplier Novus Biologicals Supplier No. NBP326672100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CDC25B Monoclonal antibody specifically detects CDC25B in Human samples. It is validated for ELISA

Specifications

Antigen CDC25B
Applications ELISA
Classification Monoclonal
Clone 3D6
Conjugate Unconjugated
Dilution ELISA
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias CDC25HU2, cell division cycle 25 homolog B (S. cerevisiae), cell division cycle 25 homolog B (S. pombe), cell division cycle 25B, Dual specificity phosphatase Cdc25B, EC 3.1.3.48, M-phase inducer phosphatase 2
Host Species Rabbit
Immunogen A synthesized peptide derived from Human CDC25B [UniProt P30305]
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 994
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.