Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDC25B Antibody (3D6), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | CDC25B |
---|---|
Clone | 3D6 |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Description
CDC25B Monoclonal antibody specifically detects CDC25B in Human samples. It is validated for ELISASpecifications
CDC25B | |
ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CDC25HU2, cell division cycle 25 homolog B (S. cerevisiae), cell division cycle 25 homolog B (S. pombe), cell division cycle 25B, Dual specificity phosphatase Cdc25B, EC 3.1.3.48, M-phase inducer phosphatase 2 | |
A synthesized peptide derived from Human CDC25B [UniProt P30305] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
3D6 | |
ELISA | |
Unconjugated | |
Rabbit | |
Cancer, Cell Cycle and Replication, Phospho Specific | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
994 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title