Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDC25B Antibody, Novus Biologicals™
SDP

Catalog No. NBP215826 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP215826 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP215826 Supplier Novus Biologicals Supplier No. NBP215826
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CDC25B Polyclonal specifically detects CDC25B in Human samples. It is validated for Western Blot.

Specifications

Antigen CDC25B
Applications Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000
Formulation PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. P30305
Gene Alias CDC25HU2, cell division cycle 25 homolog B (S. cerevisiae), cell division cycle 25 homolog B (S. pombe), cell division cycle 25B, Dual specificity phosphatase Cdc25B, EC 3.1.3.48, M-phase inducer phosphatase 2
Gene Symbols CDC25B
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human CDC25B. The exact sequence is proprietary.
Molecular Weight of Antigen 65 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 994
Test Specificity Human. Expected to cross react based on sequence identity: Mouse (93%), Pig (93%), Rat (92%), Bovine (93%).
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.