Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDC25B (p Ser323) Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
SDP

Catalog No. NBP254756 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP254756 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP254756 Supplier Novus Biologicals Supplier No. NBP254756

Rabbit Polyclonal Antibody

Specifications

Antigen CDC25B (p Ser323)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000
Formulation Liquid in 0.42% Potassium phosphate, 0.87% Sodium chloride, pH 7.3, and 30% glycerol. with 0.01% Sodium Azide
Gene Alias CDC25HU2, cell division cycle 25 homolog B (S. cerevisiae), cell division cycle 25 homolog B (S. pombe), cell division cycle 25B, Dual specificity phosphatase Cdc25B, EC 3.1.3.48, M-phase inducer phosphatase 2
Gene Symbols CDC25B
Host Species Rabbit
Immunogen KLH-conjugated synthetic peptide encompassing a sequence within the center region of human CDC25B (pS323). The exact sequence is proprietary.
Purification Method Immunogen affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 994
Target Species Human, Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.