Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDC25C, p Ser216 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100818410.025ML
Description
CDC25C Polyclonal specifically detects CDC25C in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
CDC25C (p Ser216) | |
Polyclonal | |
Unconjugated | |
phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, and 50% glycerol. with 0.02% Sodium Azide | |
CDC 25, CDC25, cell division cycle 25 homolog C (S. cerevisiae), cell division cycle 25 homolog C (S. pombe), cell division cycle 25C, Dual specificity phosphatase Cdc25C, EC 3.1.3.48, mitosis inducer CDC25, M-phase inducer phosphatase 3, phosphotyrosine phosphatase | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
cdc25C (specific to Phospho-Ser216) detects endogenous levels of cdc25C only when phosphorylated at serine 216. | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunohistochemistry, Immunofluorescence | |
1.0 mg/mL | |
Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:100-1:200, Immunohistochemistry-Paraffin 1:50-1:100 | |
P30307 | |
CDC25C | |
The antiserum was produced against synthesized phosphopeptide derived from human cdc25C around the phosphorylation site of serine 216 (S-P-SP-M-P). | |
0.025 mL | |
Phospho Specific | |
995 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction