Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDC25C, p Ser216 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$243.50 - $402.00
Specifications
Antigen | CDC25C (p Ser216) |
---|---|
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:100-1:200, Immunohistochemistry-Paraffin 1:50-1:100 |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunohistochemistry, Immunofluorescence |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017827
![]() |
Novus Biologicals
NB100818410.025ML |
0.025 mL |
Each for $243.50
|
|
|||||
NB10081841
![]() |
Novus Biologicals
NB10081841 |
0.05 mL |
Each for $402.00
|
|
|||||
Description
CDC25C Polyclonal specifically detects CDC25C in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
CDC25C (p Ser216) | |
Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:100-1:200, Immunohistochemistry-Paraffin 1:50-1:100 | |
Polyclonal | |
Rabbit | |
Phospho Specific | |
P30307 | |
995 | |
The antiserum was produced against synthesized phosphopeptide derived from human cdc25C around the phosphorylation site of serine 216 (S-P-SP-M-P). | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
1.0 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Human | |
CDC 25, CDC25, cell division cycle 25 homolog C (S. cerevisiae), cell division cycle 25 homolog C (S. pombe), cell division cycle 25C, Dual specificity phosphatase Cdc25C, EC 3.1.3.48, mitosis inducer CDC25, M-phase inducer phosphatase 3, phosphotyrosine phosphatase | |
CDC25C | |
IgG | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title