Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDC26 Antibody, Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB042614 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB042614 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB042614 Supplier Novus Biologicals Supplier No. NBP297346AF750
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CDC26 Polyclonal specifically detects CDC26 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen CDC26
Applications Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 750
Dilution Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias ANAPC12chromosome 9 open reading frame 17, Anaphase-promoting complex subunit 12, anaphase-promoting complex subunit CDC26, APC12anaphase promoting complex subunit 12, C9orf17, CDC26 subunit of anaphase promoting complex, cell division cycle 26, cell division cycle 26 homolog (S. cerevisiae), Cell division cycle protein 26 homolog
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human CDC26 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 246184
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.