Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cdc27 Antibody (AF 3.1), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB1002606C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002606C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002606C Supplier Novus Biologicals Supplier No. NB1002606C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cdc27 Monoclonal specifically detects Cdc27 in Mammalia, Xenopus species samples. It is validated for CyTOF, ELISA, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot.

Specifications

Antigen Cdc27
Applications CyTOF, ELISA, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone AF 3.1
Conjugate DyLight 650
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias ANAPC3CDC27 homolog, anaphase promoting complex subunit 3, Anaphase-promoting complex subunit 3, anaphase-promoting complex, protein 3, APC 3, APC3cell division cycle 27, CDC27Hs, cell division cycle 27 homolog (S. cerevisiae), cell division cycle protein 27 homolog, D0S1430E, D17S978E, HNUC, H-NUC, NUC2, nuc2 homolog
Gene Symbols CDC27
Host Species Mouse
Immunogen A peptide corresponding to the C-terminal 10 residues of human cdc27 coupled to KLH.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 996
Target Species Mammalia, Xenopus species
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.