Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cdc27 Rabbit anti-Human, Clone: SD85-02, Novus Biologicals™
SDP

Catalog No. NBP267828 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP267828 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP267828 Supplier Novus Biologicals Supplier No. NBP267828

Rabbit Monoclonal Antibody

Cdc27 Monoclonal specifically detects Cdc27 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen Cdc27
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SD85-02
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:5000, Immunohistochemistry-Paraffin 1:200
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias ANAPC3CDC27 homolog, anaphase promoting complex subunit 3, Anaphase-promoting complex subunit 3, anaphase-promoting complex, protein 3, APC 3, APC3cell division cycle 27, CDC27Hs, cell division cycle 27 homolog (S. cerevisiae), cell division cycle protein 27 homolog, D0S1430E, D17S978E, HNUC, H-NUC, NUC2, nuc2 homolog
Gene Symbols CDC27
Host Species Rabbit
Immunogen Recombinant protein within Human Cdc27 aa 662-824/ 824. (SwissProt: P30260 Human; SwissProt: A2A6Q5 Mouse; SwissProt: Q4V8A2 Rat)
Purification Method Protein A purified
Quantity 100 μL
Research Discipline Cell Cycle and Replication, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 996
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.