Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDC2L1 Antibody, Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB133102 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB133102 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB133102 Supplier Novus Biologicals Supplier No. NBP306170AF594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CDC2L1 Polyclonal specifically detects CDC2L1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen CDC2L1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 594
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias CDC2L1p58 CLK-1, CDC-related protein kinase p58, CDK11-p110, CDK11-p46, CDK11-p58, CDK11PITSLRE serine/threonine-protein kinase CDC2L1, cell division cycle 2-like 1 (PITSLRE proteins), Cell division cycle 2-like protein kinase 1, Cell division protein kinase 11B, cell division protein kinase 11B-like, CLK-1, cyclin-dependent kinase 11B, EC 2.7.11, EC 2.7.11.22, FLJ59152, Galactosyltransferase-associated protein kinase p58/GTA, p58, p58CDC2L1, p58CLK-1, PITSLREA, PK58
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human CDC2L1.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cell Cycle and Replication, Cellular Markers, Protein Phosphatase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 984
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.