Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDC2L2 Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB291954 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB291954 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB291954 Supplier Novus Biologicals Supplier No. NB100495PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CDC2L2 Polyclonal specifically detects CDC2L2 in Human samples. It is validated for Immunoprecipitation, Western Blot.

Specifications

Antigen CDC2L2
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate PE
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias CDK11-p110, CDK11-p58, cyclin-dependent kinase 11A, PITSLRE protein kinase beta
Host Species Rabbit
Immunogen Maps to a region between residues 50 and 100 of human Cell Division Cycle 2-like 2 using the numbering given in SwissProt entry Q9UQ88 (GeneID 985).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cell Cycle and Replication, Cellular Markers, Protein Phosphatase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 728642
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.