Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDC2L5 Antibody, Novus Biologicals™
SDP

Catalog No. NB10068268 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10068268 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10068268 Supplier Novus Biologicals Supplier No. NB10068268
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CDC2L5 Polyclonal specifically detects CDC2L5 in Human samples. It is validated for Immunoprecipitation, Western Blot.

Specifications

Antigen CDC2L5
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 ug/mg lysate
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. Q14004
Gene Alias CDC2L5, CDC2LEC 2.7.11.22, CDC2-related protein kinase 5, Cell division cycle 2-like protein kinase 5, Cell division protein kinase 13, CHEDcell division cycle 2-like 5 (cholinesterase-related cell division controller), Cholinesterase-related cell division controller, cyclin-dependent kinase 13, EC 2.7.11, EC 2.7.11.23, hCDK13, KIAA1791FLJ35215
Gene Symbols CDK13
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 1125 and 1175 of human cell division cycle 2-like 5 using the numbering given in entry NP_003709.3 (GeneID 8621).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 8621
Test Specificity Based on 100% sequence identity, this antibody is predicted to react with Panda, Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, White-tufted-ear marmoset, Chinese hamster, Naked mole rat, Small-eared galago and Northern white-cheeked gibbon.
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.