Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDC42 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP176330 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP176330 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP176330 Supplier Novus Biologicals Supplier No. NBP176330
Add to Cart
Add to Cart

Chicken Polyclonal Antibody

CDC42 Polyclonal specifically detects CDC42 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.

Specifications

Antigen CDC42
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5-1 μg/mL, ELISA 1:100-1:2000
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. NP_001782
Gene Alias CDC42Hs, cell division control protein 42 homolog, cell division cycle 42 (GTP binding protein, 25kDa), cell division cycle 42 (GTP-binding protein, 25kD), dJ224A6.1.1 (cell division cycle 42 (GTP-binding protein, 25kD)), dJ224A6.1.2 (cell division cycle 42 (GTP-binding protein, 25kD)), G25K, G25K GTP-binding protein, growth-regulating protein, GTP-binding protein, 25kD, small GTP binding protein CDC42
Gene Symbols CDC42
Host Species Chicken
Immunogen Antibody was raised against a 17 amino acid synthetic peptide near the amino terminus of human CDC42. The immunogen is located within amino acids 40 - 90 of CDC42. Amino Acid Squence: FDTAGQEDYDRLRPLSY
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 998
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgY
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.