Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDC42 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB319414 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB319414 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB319414 Supplier Novus Biologicals Supplier No. NBP298725PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CDC42 Polyclonal antibody specifically detects CDC42 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation

Specifications

Antigen CDC42
Applications Western Blot, ELISA, Immunoprecipitation
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, ELISA, Immunoprecipitation
Formulation PBS
Gene Alias CDC42Hs, cell division control protein 42 homolog, cell division cycle 42 (GTP binding protein, 25kDa), cell division cycle 42 (GTP-binding protein, 25kD), dJ224A6.1.1 (cell division cycle 42 (GTP-binding protein, 25kD)), dJ224A6.1.2 (cell division cycle 42 (GTP-binding protein, 25kD)), G25K, G25K GTP-binding protein, growth-regulating protein, GTP-binding protein, 25kD, small GTP binding protein CDC42
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human CDC42 (Accession#: P60953-2; Met1-Cys188)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 998
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.