Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDC42BPB Antibody, DyLight 488, Novus Biologicals™
SDP

Catalog No. NB054617 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB054617 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB054617 Supplier Novus Biologicals Supplier No. NBP298656G
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CDC42BPB Polyclonal specifically detects CDC42BPB in Human samples. It is validated for Western Blot.

Specifications

Antigen CDC42BPB
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 488
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias CDC42 binding protein kinase beta (DMPK-like), CDC42-binding protein kinase beta, CDC42-binding protein kinase beta (DMPK-like), DMPK-like beta, EC 2.7.11, EC 2.7.11.1, FLJ37601, FLJ44730, KIAA1124DKFZp686H1431, MRCK beta, MRCKB, Myotonic dystrophy kinase-related CDC42-binding kinase beta, Myotonic dystrophy protein kinase-like beta, serine/threonine-protein kinase MRCK beta
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human CDC42BPB fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 9578
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.