Learn More
Description
Specifications
Specifications
| Antigen | CDC42EP1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS buffer, 2% sucrose |
| Gene Alias | Binder of Rho GTPases 5, Borg5, BORG5cdc42 effector protein 1, CDC42 effector protein (Rho GTPase binding) 1, CEP155 kDa bone marrow stromal/endothelial cell protein, MSE55MGC15316, serum constituent protein, Serum protein MSE55 |
| Host Species | Rabbit |
| Immunogen | The immunogen is a synthetic peptide directed towards the N-terminal region of Human CDC42EP1 (NP_689449). Peptide sequence HTMHVGRGGDVFGDTSFLSNHGGSSGSTHRSPRSFLAKKLQLVRRVGAPP |
| Purification Method | Affinity purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
