Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDC42EP3 Antibody, Novus Biologicals™
SDP

Catalog No. NB10088141 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10088141 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10088141 Supplier Novus Biologicals Supplier No. NB10088141
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CDC42EP3 Polyclonal specifically detects CDC42EP3 in Human samples. It is validated for Immunoprecipitation.

Specifications

Antigen CDC42EP3
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Immunoprecipitation 2 - 5 ug/mg lysate
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. NP_006440.2
Gene Alias Binder of Rho GTPases 2, BORG2CEP3UB1, CDC42 effector protein (Rho GTPase binding) 3, cdc42 effector protein 3, CRIB-containing BORG2 protein, FLJ46903, MSE55-related Cdc42-binding protein, MSE55-related protein
Gene Symbols CDC42EP3
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 204 and 254 of human binder of Rho GTPases 2 using the numbering given in entry NP_006440.2 (GeneID 10602).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10602
Test Specificity Based on 100% sequence identity, this antibody is predicted to react with Panda, Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, West Indian ocean coelacanth, Duckbill platypus, Tasmanian devil and Northern white-cheeked gibbon.
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.