Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDC42EP4 Antibody, Novus Biologicals™
SDP

Catalog No. NBP153150 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP153150 100 μL
NBP15315020 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP153150 Supplier Novus Biologicals Supplier No. NBP153150
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CDC42EP4 Polyclonal specifically detects CDC42EP4 in Human samples. It is validated for Western Blot.

Specifications

Antigen CDC42EP4
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q9H3Q1
Gene Alias BORG4MGC3740, CDC42 effector protein (Rho GTPase binding) 4, cdc42 effector protein 4, CEP4Binder of Rho GTPases 4, KAIA1777, MGC17125
Gene Symbols CDC42EP4
Host Species Rabbit
Immunogen Synthetic peptides corresponding to CDC42EP4(CDC42 effector protein (Rho GTPase binding) 4) The peptide sequence was selected from the N terminal of CDC42EP4. Peptide sequence SSSKRSLLSRKFRGSKRSQSVTRGEREQRDMLGSLRDSALFVKNAMSLPQ.
Molecular Weight of Antigen 38 kDa
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23580
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.