Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cdc6 Antibody (cdc6 9H8/5), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. N102684A350 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N102684A350 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N102684A350 Supplier Novus Biologicals Supplier No. NB1002684AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cdc6 Monoclonal specifically detects Cdc6 in Yeast samples. It is validated for Immunohistochemistry, Immunoprecipitation, Western Blot.

Specifications

Antigen Cdc6
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone cdc6 9H8/5
Conjugate Alexa Fluor 350
Dilution Western Blot, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias CDC18LHsCDC18, Cdc18-related protein, CDC6 (cell division cycle 6, S. cerevisiae) homolog, CDC6 cell division cycle 6 homolog, CDC6 cell division cycle 6 homolog (S. cerevisiae), CDC6-related protein, cell division control protein 6 homolog, cell division cycle 6 homolog (S. cerevisiae), cell division cycle 6 protein, HsCdc18, HsCDC6, p62(cdc6)
Gene Symbols CDC6
Host Species Mouse
Immunogen Cdc6 from S. cerevisiae.
Purification Method Protein A purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 990
Target Species Yeast
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.