Learn More
CDC6 Mouse anti-Human, Mouse, Rat, Clone: 6F12D2, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | CDC6 |
---|---|
Clone | 6F12D2 |
Concentration | 0.75 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6464
![]() ![]() |
Proteintech Group Inc
660211IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This antibody is a mouse monoclonal antibody raised against residues near the C terminus of human CDC6.
The protein encoded by this gene is highly similar to Saccharomyces cerevisiae Cdc6, a protein essential for the initiation of DNA replication. This protein functions as a regulator at the early steps of DNA replication. It localizes in cell nucleus during cell cyle G1, but translocates to the cytoplasm at the start of S phase. The subcellular translocation of this protein during cell cyle is regulated through its phosphorylation by Cdks. Transcription of this protein was reported to be regulated in response to mitogenic signals through transcriptional control mechanism involving E2F proteins.Specifications
CDC6 | |
0.75 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Cdc18 related protein, CDC18L, CDC6, CDC6 related protein, HsCDC18, HsCDC6, p62(cdc6) | |
Cdc6 | |
IgG1 | |
Protein G | |
Antibody |
6F12D2 | |
Immunocytochemistry, Immunofluorescence, Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat | |
O89033, Q99741 | |
23834, 360621, 990 | |
CDC6 Fusion Protein Ag17958 | |
Primary | |
-20°C | |
CDC6 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.