Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDC73/HRPT2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$343.50 - $573.00
Specifications
| Antigen | CDC73/HRPT2 |
|---|---|
| Dilution | Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
CDC73/HRPT2 Polyclonal antibody specifically detects CDC73/HRPT2 in Human samples. It is validated for ImmunofluorescenceSpecifications
| CDC73/HRPT2 | |
| Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cancer, Tumor Suppressors | |
| PBS, pH 7.2, 40% glycerol | |
| 79577 | |
| IgG | |
| Affinity purified |
| Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| C1orf28hyperparathyroidism 2 (with jaw tumor), cell division cycle 73, Paf1/RNA polymerase II complex component, homolog (S.cerevisiae), Cell division cycle protein 73 homolog, chromosome 1 open reading frame 28, HPTJT, HRPT2FLJ23316, Hyperparathyroidism 2 protein, HYX, parafibromin | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: WDRVVAVFVQGPAWQFKGWPWLLPDGSPVDIFAKIKAFHLKYDEVRLDPNVQKWDVTVLELSYHKRHLDRPVFLRFWETLDRYMVKHKSHL | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title