Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDC73 Mouse anti-Human, Clone: 2H1-F10-B9, MilliporeSigma™

Mouse Monoclonal Antibody

Supplier:  MilliporeSigma MABC176825UG

 View more versions of this product

Catalog No. MABC176825


Only null left
Add to Cart

Description

Description

CDC73 Monoclonal antibody specifically detects CDC73 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry, Western Blotting.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CDC73
Monoclonal
1.0 mg/mL
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
KLH-conjugated linear peptide corresponding to 13 amino acids from the N-terminal region of human CDC73 (Parafibromin).
25 μg
Primary
Stable for 1 year at +2°C to +8°C from date of receipt.
IgG1 κ
Immunocytochemistry, Immunohistochemistry, Western Blot
2H1-F10-B9
Unconjugated
Mouse
Protein G purified
RUO
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.