Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDCA3 Antibody, Novus Biologicals™
SDP

Catalog No. NB10074621 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10074621 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10074621 Supplier Novus Biologicals Supplier No. NB10074621

Rabbit Polyclonal Antibody

CDCA3 Polyclonal specifically detects CDCA3 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative).

Specifications

Antigen CDCA3
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Immunoprecipitation 2-5ug/mg Lysate, Western Blot (Negative)
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. NP_112589.1
Gene Alias C8, cell division cycle associated 3, FBXO52, Gene-rich cluster protein C8, GRCC8cell division cycle-associated protein 3, MGC2577, TOME1, TOME-1gene rich cluster, C8, trigger of mitotic entry 1, Trigger of mitotic entry protein 1
Gene Symbols CDCA3
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 200 and 230 of human cell division cycle associated 3 (trigger of mitotic entry 1) using the numbering given in entry NP_112589.1 (GeneID 83461).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 83461
Test Specificity Human.Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, Crab-eating macaque, Chinese hamster, Little brown bat and Northern whitecheeked gibbon.
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.