Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDCA7L Antibody, Novus Biologicals™
SDP

Catalog No. NBP224472SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224472SS 0.025 mg
NBP224472 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224472SS Supplier Novus Biologicals Supplier No. NBP224472SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CDCA7L Polyclonal specifically detects CDCA7L in Human, Primate samples. It is validated for Western Blot.

Specifications

Antigen CDCA7L
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2-5 ug/ml
Formulation 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Alias cell division cycle associated 7-like, DKFZp762L0311, JPO2cell division cycle-associated 7-like protein, Protein JPO2, R1HR1, RAM2, Transcription factor RAM2
Gene Symbols CDCA7L
Host Species Rabbit
Immunogen A portion of amino acid 170 to 220 of human CDCA7L/RAM2 was used as immunogen.
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55536
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human and chimpanzee, 94% homologous in monkey, 89% homologous in mouse, rat and horse, and 84% homologous in cow.
Target Species Human, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.