Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDCP1 Antibody (309116), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB364548 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB364548 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB364548 Supplier Novus Biologicals Supplier No. FAB26663J
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CDCP1 Monoclonal antibody specifically detects CDCP1 in Human samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen CDCP1
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 309116
Conjugate Janelia Fluor 646
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD318, CD318 antigen, CUB domain containing protein 1, CUB domain-containing protein 1, Membrane glycoprotein gp140, SIMA135TRASK, Subtractive immunization M plus HEp3-associated 135 kDa protein, Transmembrane and associated with src kinases
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human CDCP1, Ala33-Leu666, Accession # NP_073753
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64866
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.