Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDCP1 Antibody (30D4), Novus Biologicals™
SDP

Catalog No. NB297643 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB297643 100 μL
NB297642 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB297643 Supplier Novus Biologicals Supplier No. NBP326667100UL
Add to Cart
Add to Cart

Human Monoclonal Antibody

CDCP1 Monoclonal antibody specifically detects CDCP1 in Human, Mouse, Cynomolgus Monkey samples. It is validated for ELISA

Specifications

Antigen CDCP1
Applications ELISA
Classification Monoclonal
Clone 30D4
Conjugate Unconjugated
Dilution ELISA
Formulation 50% Glycerol, 0.01M PBS, PH 7.4
Gene Alias CD318, CD318 antigen, CUB domain containing protein 1, CUB domain-containing protein 1, Membrane glycoprotein gp140, SIMA135TRASK, Subtractive immunization M plus HEp3-associated 135 kDa protein, Transmembrane and associated with src kinases
Host Species Human
Immunogen Recombinant Human CDCP1 protein [UniProt Q9H5V8]
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 64866
Target Species Human, Mouse, Cynomolgus Monkey
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.