Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDCP1 Antibody (30D4), Novus Biologicals™

Human Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | CDCP1 |
---|---|
Clone | 30D4 |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Description
CDCP1 Monoclonal antibody specifically detects CDCP1 in Human, Mouse, Cynomolgus Monkey samples. It is validated for ELISASpecifications
CDCP1 | |
ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Cynomolgus Monkey | |
CD318, CD318 antigen, CUB domain containing protein 1, CUB domain-containing protein 1, Membrane glycoprotein gp140, SIMA135TRASK, Subtractive immunization M plus HEp3-associated 135 kDa protein, Transmembrane and associated with src kinases | |
Recombinant Human CDCP1 protein [UniProt Q9H5V8] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
30D4 | |
ELISA | |
Unconjugated | |
Human | |
Phospho Specific | |
50% Glycerol, 0.01M PBS, PH 7.4 | |
64866 | |
IgG1 | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title