Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDCP1 Antibody (OTI2C1), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP27216A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP27216A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP27216A32 Supplier Novus Biologicals Supplier No. NBP272156AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CDCP1 Monoclonal specifically detects CDCP1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen CDCP1
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI2C1
Conjugate Alexa Fluor 532
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias CD318, CD318 antigen, CUB domain containing protein 1, CUB domain-containing protein 1, Membrane glycoprotein gp140, SIMA135TRASK, Subtractive immunization M plus HEp3-associated 135 kDa protein, Transmembrane and associated with src kinases
Gene Symbols CDCP1
Host Species Mouse
Immunogen Full length recombinant protein of human CDCP1 (NP_073753) produced in HEK293T cells.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 64866
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.