Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDH12 Monoclonal Antibody (01), Invitrogen™

Mouse Monoclonal Antibody
$285.50 - $493.00
Specifications
Antigen | CDH12 |
---|---|
Clone | 01 |
Concentration | 1 mg/mL |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA543513
|
Invitrogen™
MA543513 |
50 μL |
Each for $285.50
|
|
|||||
PIMA542983
![]() |
Invitrogen™
MA542983 |
100 μL |
Each for $493.00
|
|
|||||
Description
Specificity: Human Cadherin-12/CDH12
CDH12 is a type II classical cadherin from the cadherin superfamily of integral membrane proteins that mediate calcium-dependent cell-cell adhesion. Mature cadherin proteins are composed of a large N-terminal extracellular domain, a single membrane-spanning domain, and a small, highly conserved C-terminal cytoplasmic domain. Type II (atypical) cadherins are defined based on their lack of a HAV cell adhesion recognition sequence specific to type I cadherins. This particular cadherin appears to be expressed specifically in the brain and its temporal pattern of expression would be consistent with a role during a critical period of neuronal development, perhaps specifically during synaptogenesis.Specifications
CDH12 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with no preservative | |
Brain cadherin; brain-cadherin; Br-cadherin; Cadherin; cadherin 12; cadherin 12, type 2 (N-cadherin 2); cadherin, neural type, 2; cadherin-12; CDH12; CDHB; N-cadherin 2; neural type cadherin 2; neuronal cadherin 2; RGD1566350 | |
CDH12 | |
IgG1 | |
Protein A | |
Antibody |
01 | |
ELISA | |
Unconjugated | |
Mouse | |
Human | |
P55289 | |
1010 | |
Recombinant Human Cadherin-12/CDH12 Protein. | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
CDH12 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title