Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDH17 Mouse anti-Human, Unconjugated, Clone: Lic5, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABT1525100UG
Description
CDH17 Monoclonal antibody specifically detects CDH17 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Inhibition, Western Blotting.
Specifications
CDH17 | |
Monoclonal | |
Unconjugated | |
Intestinal peptide-associated transporter HPT-1;Liver-intestine cadherin;LI-cadherin | |
Mouse | |
92.22 kDa calculated. | |
100 μg | |
Primary | |
Human | |
Purified |
Immunocytochemistry, Immunohistochemistry (Paraffin), Inhibition Assays, Western Blot | |
Lic5 | |
NP_001138135 | |
CDH17 | |
Recombinant fragment corresponding to extracellular domains 1 and 2 of human Cadherin-17 (CDH17). | |
Protein G Purified | |
Cell Structure | |
Clone Lic5 is a mouse monoclonal antibody that specifically detects human Cadherin-17. It targets an epitope within the extracellular domain. | |
Stable for 1 year at -20°C from date of receipt. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction