Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDH17 Mouse anti-Human, Unconjugated, Clone: Lic5, MilliporeSigma™
Mouse Monoclonal Antibody
$182.31 - $457.74
Specifications
Antigen | CDH17 |
---|---|
Clone | Lic5 |
Applications | Immunocytochemistry, Immunohistochemistry (Paraffin), Inhibition Assays, Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABT15252MI
|
MilliporeSigma
MABT152525UG |
25 μg |
Each for $182.31
|
|
|||||
MABT15251MI
|
MilliporeSigma
MABT1525100UG |
100 μg |
Each for $457.74
|
|
|||||
Description
CDH17 Monoclonal antibody specifically detects CDH17 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Inhibition, Western Blotting.Specifications
CDH17 | |
Immunocytochemistry, Immunohistochemistry (Paraffin), Inhibition Assays, Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Intestinal peptide-associated transporter HPT-1;Liver-intestine cadherin;LI-cadherin | |
Recombinant fragment corresponding to extracellular domains 1 and 2 of human Cadherin-17 (CDH17). | |
Primary | |
Stable for 1 year at -20°C from date of receipt. | |
92.22 kDa calculated. |
Lic5 | |
Monoclonal | |
Purified | |
Cell Structure | |
NP_001138135 | |
CDH17 | |
IgG2b κ | |
Protein G Purified | |
Clone Lic5 is a mouse monoclonal antibody that specifically detects human Cadherin-17. It targets an epitope within the extracellular domain. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title