Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDIP Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB356851 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB356851 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB356851 Supplier Novus Biologicals Supplier No. NBP176996PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CDIP Polyclonal antibody specifically detects CDIP in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen CDIP
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias CDIP, cell death inducing protein, Cell death-inducing protein, chromosome 16 open reading frame 5, I1, LITAF-like protein, Transmembrane protein I1
Host Species Rabbit
Immunogen Antibody was raised against a 19 amino acid synthetic peptide near the center of human CDIP. The immunogen is located within amino acids 70 - 120 of CDIP. Amino Acid Squence: YPPPGPHPPMGYYPPGPYT
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 29965
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.