Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDK2 Antibody (C15.2), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB1002686AF Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002686AF 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002686AF Supplier Novus Biologicals Supplier No. NB1002686AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CDK2 Monoclonal specifically detects CDK2 in Xenopus species samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Western Blot.

Specifications

Antigen CDK2
Applications ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone C15.2
Conjugate Alexa Fluor 405
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias cdc2-related protein kinase, cell devision kinase 2, Cell division protein kinase 2, cyclin-dependent kinase 2, EC 2.7.11, EC 2.7.11.22, p33 protein kinase, p33(CDK2)
Gene Symbols CDK2
Host Species Mouse
Immunogen Xenopus Cdk2
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cell Cycle and Replication, Ovarian Carcinoma Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 1017
Target Species Xenopus species
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.